Assay Details
| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013871.3 | NP_038899.1 | 8 - 9 | 918 | 84 | |
| GenBank mRNA | AK011286.1 | - | 7 - 8 | 958 | 84 | |
| BC021640.1 | - | 8 - 9 | 836 | 3979667 | 84 | |
| Y13439.1 | - | 8 - 9 | 854 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk12
Genomic Map