Assay Details
| Gene Symbol: Hnf4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013920.2 | NP_038948.1 | 1 - 2 | 306 | 98 | |
| GenBank mRNA | AJ242626.1 | - | 1 - 2 | 258 | 98 | |
| AK143816.1 | - | 1 - 2 | 307 | 98 | ||
| BC125519.1 | - | 1 - 2 | 128 | 40130034 | 98 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ242626.1,AK143816.1,BC125519.1,NM_013920.2 transcripts.
Set Membership: |
|

Hnf4g
Genomic Map