Assay Details
| Gene Symbol: Hnf4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013920.2 | NP_038948.1 | 4 - 5 | 673 | 96 | |
| GenBank mRNA | AJ242626.1 | - | 4 - 5 | 625 | 96 | |
| AK143816.1 | - | 4 - 5 | 674 | 96 | ||
| BC125519.1 | - | 4 - 5 | 495 | 40130034 | 96 | |
More Information
Set Membership: |
|

Hnf4g
Genomic Map