Assay Details
| Gene Symbol: Tmprss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015775.2 | NP_056590.2 | 9 - 10 | 1118 | 94 | |
| GenBank mRNA | AF113596.1 | - | 9 - 10 | 988 | 94 | |
| AF199362.1 | - | 9 - 10 | 999 | 94 | ||
| AF243500.1 | - | 9 - 10 | 988 | 94 | ||
| AK141350.1 | - | 9 - 10 | 1093 | 94 | ||
| AK146047.1 | - | 9 - 10 | 1002 | 94 | ||
| BC038393.1 | - | 9 - 10 | 957 | 2648878 | 94 | |
| BC054348.1 | - | 9 - 10 | 1118 | 2648889 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss2
Genomic Map