Assay Details
| Gene Symbol: Chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016681.3 | NP_057890.1 | 10 - 11 | 1147 | 125 | |
| GenBank mRNA | AF086905.1 | - | 9 - 10 | 1108 | 125 | |
| AK044913.1 | - | 10 - 11 | 1148 | 125 | ||
| AK170235.1 | - | 9 - 10 | 1091 | 125 | ||
| BC056617.1 | - | 10 - 11 | 1171 | 4977960 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chek2
Genomic Map