Assay Details
| Gene Symbol: Chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016681.3 | NP_057890.1 | 11 - 12 | 1305 | 72 | |
| GenBank mRNA | AF086905.1 | - | 10 - 11 | 1266 | 72 | |
| AK044913.1 | - | 11 - 12 | 1306 | 72 | ||
| AK170235.1 | - | 10 - 11 | 1249 | 72 | ||
| BC056617.1 | - | 11 - 12 | 1329 | 4977960 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chek2
Genomic Map