Assay Details
| Gene Symbol: Clcn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243762.1 | NP_001230691.1 | 11 - 12 | 2144 | 75 | |
| NM_016691.4 | NP_057900.3 | 9 - 10 | 1762 | 75 | ||
| GenBank mRNA | AF134117.1 | - | 9 - 10 | 1762 | 75 | |
| AK052990.1 | - | 9 - 10 | 1742 | 75 | ||
| AK142227.1 | - | 9 - 10 | 1694 | 75 | ||
| AK145763.1 | - | 9 - 10 | 1731 | 75 | ||
| BC036347.1 | - | 9 - 10 | 1734 | 3989812 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcn5
Genomic Map