Assay Details
| Gene Symbol: Mybph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016749.2 | NP_058029.2 | 5 - 6 | 871 | 73 | |
| GenBank mRNA | BC116684.1 | - | 5 - 6 | 838 | 40106746 | 73 |
| BC118050.1 | - | 5 - 6 | 838 | 40106744 | 73 | |
| CB587371.1 | - | 1 - 2 | 118 | 73 | ||
| U68267.1 | - | 5 - 6 | 865 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mybph
Genomic Map