Assay Details
| Gene Symbol: Rgs14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016758.3 | NP_058038.2 | 13 - 14 | 1548 | 70 | |
| GenBank mRNA | AK002891.1 | - | 8 - 9 | 1037 | 70 | |
| BC030321.1 | - | 13 - 14 | 1520 | 5374531 | 70 | |
| U85055.1 | - | 13 - 14 | 1388 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs14
Genomic Map