Assay Details
| Gene Symbol: Itgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016780.2 | NP_058060.2 | 6 - 7 | 1027 | 64 | |
| GenBank mRNA | AF026509.1 | - | 6 - 7 | 971 | 64 | |
| AK156518.1 | - | 2 - 3 | 284 | 64 | ||
| AK157958.1 | - | 6 - 7 | 973 | 64 | ||
| AK198913.1 | - | 88 | 64 | |||
| BC125518.1 | - | 6 - 7 | 1027 | 40130033 | 64 | |
| BC125520.1 | - | 6 - 7 | 1027 | 40130035 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb3
Genomic Map