Assay Details
| Gene Symbol: Itgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016780.2 | NP_058060.2 | 12 - 13 | 2097 | 71 | |
| GenBank mRNA | AF026509.1 | - | 12 - 13 | 2041 | 71 | |
| AK156518.1 | - | 8 - 9 | 1355 | 71 | ||
| AK157958.1 | - | 12 - 13 | 2043 | 71 | ||
| BC125518.1 | - | 12 - 13 | 2097 | 40130033 | 71 | |
| BC125520.1 | - | 12 - 13 | 2097 | 40130035 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb3
Genomic Map