Assay Details
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016797.4 | NP_058077.2 | 4 - 5 | 723 | 105 | |
| GenBank mRNA | AB019212.1 | - | 4 - 5 | 393 | 105 | |
| AF056323.1 | - | 4 - 5 | 335 | 105 | ||
| AK002867.1 | - | 4 - 5 | 392 | 105 | ||
| AK013869.1 | - | 4 - 5 | 377 | 105 | ||
| AK082927.1 | - | 4 - 5 | 363 | 105 | ||
| AK142742.1 | - | 370 | 105 | |||
| AK158923.1 | - | 4 - 5 | 724 | 105 | ||
| AK166822.1 | - | 4 - 5 | 393 | 105 | ||
| AK169822.1 | - | 4 - 5 | 396 | 105 | ||
| AK186903.1 | - | 2 - 3 | 113 | 106 | ||
| BC132123.1 | - | 3 - 4 | 273 | 40130400 | 105 | |
| BC132125.1 | - | 3 - 4 | 273 | 40130402 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stx7
Genomic Map