Assay Details
| Gene Symbol: Dgka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016811.2 | NP_058091.2 | 2 - 3 | 399 | 73 | |
| GenBank mRNA | AF085219.1 | - | 1 - 2 | 85 | 73 | |
| BC006713.1 | - | 2 - 3 | 399 | 3584592 | 73 | |
| JN832677.1 | - | 3 - 4 | 431 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgka
Genomic Map