Assay Details
| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276704.1 | NP_001263633.1 | 6 - 7 | 751 | 66 | |
| NM_016813.2 | NP_058093.2 | 6 - 7 | 765 | 66 | ||
| GenBank mRNA | AF093140.1 | - | 6 - 7 | 741 | 66 | |
| AK077644.1 | - | 6 - 7 | 722 | 66 | ||
| AK086527.1 | - | 6 - 7 | 762 | 66 | ||
| AK088634.1 | - | 6 - 7 | 722 | 66 | ||
| AK133883.1 | - | 6 - 7 | 754 | 66 | ||
| AK143640.1 | - | 752 | 66 | |||
| AK167518.1 | - | 6 - 7 | 753 | 66 | ||
| BC005594.1 | - | 6 - 7 | 765 | 3710546 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nxf1
Genomic Map