Assay Details
| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276704.1 | NP_001263633.1 | 11 - 12 | 3003 | 65 | |
| NM_016813.2 | NP_058093.2 | 12 - 13 | 1252 | 65 | ||
| GenBank mRNA | AF093140.1 | - | 12 - 13 | 1228 | 65 | |
| AK052937.1 | - | 2 - 3 | 419 | 65 | ||
| AK077644.1 | - | 12 - 13 | 1209 | 65 | ||
| AK079583.1 | - | 12 - 13 | 1241 | 65 | ||
| AK086527.1 | - | 11 - 12 | 3014 | 65 | ||
| AK088634.1 | - | 11 - 12 | 2974 | 65 | ||
| AK133883.1 | - | 12 - 13 | 1241 | 65 | ||
| AK143640.1 | - | 3004 | 65 | |||
| AK167518.1 | - | 12 - 13 | 1240 | 65 | ||
| AK191466.1 | - | 3 - 4 | 208 | 65 | ||
| BC005594.1 | - | 12 - 13 | 1252 | 3710546 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nxf1
Genomic Map