Assay Details
| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276704.1 | NP_001263633.1 | 12 - 13 | 3054 | 77 | |
| NM_016813.2 | NP_058093.2 | 13 - 14 | 1303 | 77 | ||
| GenBank mRNA | AF093140.1 | - | 13 - 14 | 1279 | 77 | |
| AK077644.1 | - | 13 - 14 | 1260 | 77 | ||
| AK079583.1 | - | 13 - 14 | 1292 | 77 | ||
| AK086527.1 | - | 12 - 13 | 3065 | 77 | ||
| AK088634.1 | - | 12 - 13 | 3025 | 77 | ||
| AK133883.1 | - | 13 - 14 | 1292 | 77 | ||
| AK143640.1 | - | 3055 | 77 | |||
| AK167518.1 | - | 13 - 14 | 1291 | 77 | ||
| BC005594.1 | - | 13 - 14 | 1303 | 3710546 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nxf1
Genomic Map