Assay Details
| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276704.1 | NP_001263633.1 | 13 - 14 | 3157 | 94 | |
| NM_016813.2 | NP_058093.2 | 14 - 15 | 1406 | 94 | ||
| GenBank mRNA | AF093140.1 | - | 14 - 15 | 1382 | 94 | |
| AK052937.1 | - | 4 - 5 | 573 | 94 | ||
| AK077644.1 | - | 14 - 15 | 1363 | 94 | ||
| AK079583.1 | - | 14 - 15 | 1395 | 94 | ||
| AK086527.1 | - | 13 - 14 | 3168 | 94 | ||
| AK088634.1 | - | 13 - 14 | 3128 | 94 | ||
| AK133883.1 | - | 14 - 15 | 1395 | 94 | ||
| AK143640.1 | - | 3158 | 94 | |||
| AK167518.1 | - | 14 - 15 | 1394 | 94 | ||
| BC005594.1 | - | 14 - 15 | 1406 | 3710546 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nxf1
Genomic Map