Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500022726.mm.1 |
TC0500002569.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500022726.mm.1 |
NM_001122756.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022726.mm.1 |
NM_001122756.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022726.mm.1 |
NM_016869.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022726.mm.1 |
NM_016869.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022725.mm.1 |
TC0500002569.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500022725.mm.1 |
NM_001122756.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022725.mm.1 |
NM_001122756.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022725.mm.1 |
NM_016869.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022725.mm.1 |
NM_016869.3 |
Refseq |