| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100036150.mm.1 |
NM_016896.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036150.mm.1 |
NM_016896.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036150.mm.1 |
TC1100003870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100036151.mm.1 |
NM_016896.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036151.mm.1 |
NM_016896.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036151.mm.1 |
TC1100003870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100036149.mm.1 |
NM_016896.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036149.mm.1 |
NM_016896.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036149.mm.1 |
TC1100003870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100036147.mm.1 |
NM_016896.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036147.mm.1 |
NM_016896.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036147.mm.1 |
TC1100003870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100036155.mm.1 |
NM_016896.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036155.mm.1 |
NM_016896.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100036155.mm.1 |
TC1100003870.mm.1 |
MTA TC |