Assay Details
| Gene Symbol: Rab25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016899.4 | NP_058595.2 | 1 - 2 | 254 | 93 | |
| GenBank mRNA | AF119675.1 | - | 1 - 2 | 190 | 93 | |
| AK003260.1 | - | 1 - 2 | 255 | 93 | ||
| BC006624.1 | - | 1 - 2 | 240 | 3968810 | 93 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF119675.1,AK003260.1,BC006624.1,NM_016899.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
epithelial cell morphogenesis
transport
intracellular protein transport
exocytosis
small GTPase mediated signal transduction
positive regulation of cell proliferation
positive regulation of epithelial cell migration
protein transport
pseudopodium organization
melanosome transport
Rab protein signal transduction
regulation of vesicle-mediated transport
nucleotide binding
GTPase activity
protein binding
GTP binding
myosin V binding
epithelial cell morphogenesis
transport
intracellular protein transport
exocytosis
small GTPase mediated signal transduction
positive regulation of cell proliferation
positive regulation of epithelial cell migration
protein transport
pseudopodium organization
melanosome transport
Rab protein signal transduction
regulation of vesicle-mediated transport
nucleotide binding
GTPase activity
protein binding
GTP binding
myosin V binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149577 | NM_016899.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149577 | AB232620.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149577 | AF119675.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149577 | AK003260.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149577 | BC006624.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149577 | NM_016899.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149577 | AB232620.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149577 | AF119675.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149577 | AK003260.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P149577 | BC006624.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815836 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158988 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904357 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81585 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107808 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149577 | NM_016899.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149577 | AB232620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149577 | AF119675.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149577 | AK003260.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149577 | BC006624.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149577 | NM_016899.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149577 | AB232620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149577 | AF119675.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149577 | AK003260.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P149577 | BC006624.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P149577 | NM_016899.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P149577 | AB232620.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P149577 | AF119675.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P149577 | AK003260.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P149577 | BC006624.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P149577 | NM_016899.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P149577 | AB232620.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P149577 | AF119675.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P149577 | AK003260.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P149577 | BC006624.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22314433 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107808 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04981189 | - | - |
| Whole Mouse Genome G4122A | A_52_P149577 | NM_016899.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149577 | AB232620.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149577 | AF119675.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149577 | AK003260.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149577 | BC006624.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149577 | NM_016899.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149577 | AB232620.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149577 | AF119675.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149577 | AK003260.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P149577 | BC006624.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110282 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107808 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02387104 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02387107 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02387106 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02798442 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02387101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22314361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02384077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02513384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02387104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22314407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03168177 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499413 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499415 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499417 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499414 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499416 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417738_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499412 | - | - |
| Mouse Genome 430 2.0 Array | 1417738_at | - | - |
| Mouse Genome 430 2.0 Array | 1417738_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53868_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53868_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300017630.mm.1 | NM_016899.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017630.mm.1 | NM_016899.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017630.mm.1 | TC0300002316.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017632.mm.1 | NM_016899.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017632.mm.1 | NM_016899.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017632.mm.1 | TC0300002316.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417738_PM_at | - | - |
| Mouse Expression Set 430 | 1417738_at | - | - |
| Murine Genome U74v2 Set | 160731_PM_at | - | - |
| Murine Genome U74v2 Set | 160731_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417738_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417738_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417738_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417738_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1990494 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1990494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001990494 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2776087 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2776087 | - | - |
| mouse-6 v1.1 (Illumina) | scl21969.5.1_66-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1990494 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21969.5.1_66-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990494 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2781721 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2776087 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2781721 | - | - |

Rab25
Genomic Map