Assay Details
| Gene Symbol: Rab25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016899.4 | NP_058595.2 | 2 - 3 | 443 | 86 | |
| GenBank mRNA | AB232620.1 | - | 2 - 3 | 239 | 86 | |
| AF119675.1 | - | 2 - 3 | 379 | 86 | ||
| AK003260.1 | - | 2 - 3 | 444 | 86 | ||
| BC006624.1 | - | 2 - 3 | 429 | 3968810 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab25
Genomic Map