Assay Details
| Gene Symbol: Atp6v0a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243049.1 | NP_001229978.1 | 3 - 4 | 291 | 82 | |
| NM_001243050.1 | NP_001229979.1 | 4 - 5 | 472 | 82 | ||
| NM_001243051.1 | NP_001229980.1 | 4 - 5 | 472 | 82 | ||
| NM_016920.3 | NP_058616.1 | 4 - 5 | 472 | 82 | ||
| GenBank mRNA | AB022321.1 | - | 3 - 4 | 301 | 82 | |
| AF218249.1 | - | 4 - 5 | 442 | 82 | ||
| AF218250.1 | - | 3 - 4 | 295 | 82 | ||
| AF218251.1 | - | 3 - 4 | 295 | 82 | ||
| AK029316.1 | - | 4 - 5 | 444 | 82 | ||
| AK036530.1 | - | 4 - 5 | 441 | 82 | ||
| AK040343.1 | - | 4 - 5 | 380 | 82 | ||
| AK158786.1 | - | 3 - 4 | 259 | 82 | ||
| AK159114.1 | - | 4 - 5 | 408 | 82 | ||
| AK172078.1 | - | 4 - 5 | 403 | 82 | ||
| AK208028.1 | - | 1 - 2 | 50 | 82 | ||
| AK219143.1 | - | 1 - 2 | 50 | 82 | ||
| AW318708.1 | - | 4 - 5 | 402 | 82 | ||
| BC001995.1 | - | 4 - 5 | 438 | 82 | ||
| BC066839.1 | - | 4 - 5 | 437 | 30101340 | 82 | |
| BC071182.1 | - | 4 - 5 | 437 | 30101184 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0a1
Genomic Map