Assay Details
| Gene Symbol: Atp6v0a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243049.1 | NP_001229978.1 | 5 - 6 | 506 | 57 | |
| NM_001243050.1 | NP_001229979.1 | 6 - 7 | 684 | 57 | ||
| NM_001243051.1 | NP_001229980.1 | 6 - 7 | 705 | 57 | ||
| NM_016920.3 | NP_058616.1 | 6 - 7 | 684 | 57 | ||
| GenBank mRNA | AB022321.1 | - | 5 - 6 | 513 | 57 | |
| AF218249.1 | - | 6 - 7 | 654 | 57 | ||
| AF218250.1 | - | 5 - 6 | 528 | 57 | ||
| AF218251.1 | - | 5 - 6 | 507 | 57 | ||
| AK029316.1 | - | 6 - 7 | 656 | 57 | ||
| AK036530.1 | - | 6 - 7 | 652 | 57 | ||
| AK040343.1 | - | 6 - 7 | 592 | 57 | ||
| AK158786.1 | - | 5 - 6 | 474 | 57 | ||
| AK159114.1 | - | 6 - 7 | 620 | 57 | ||
| AK172078.1 | - | 6 - 7 | 614 | 57 | ||
| BC001995.1 | - | 6 - 7 | 650 | 57 | ||
| BC066839.1 | - | 6 - 7 | 649 | 30101340 | 57 | |
| BC071182.1 | - | 6 - 7 | 649 | 30101184 | 57 | |
| U13836.1 | - | 5 - 6 | 546 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0a1
Genomic Map