Assay Details
| Gene Symbol: Atp6v0a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243049.1 | NP_001229978.1 | 12 - 13 | 1469 | 107 | |
| NM_001243050.1 | NP_001229979.1 | 13 - 14 | 1647 | 107 | ||
| NM_001243051.1 | NP_001229980.1 | 13 - 14 | 1668 | 107 | ||
| NM_016920.3 | NP_058616.1 | 13 - 14 | 1647 | 107 | ||
| GenBank mRNA | AB022321.1 | - | 12 - 13 | 1476 | 107 | |
| AF218249.1 | - | 13 - 14 | 1617 | 107 | ||
| AF218250.1 | - | 12 - 13 | 1491 | 107 | ||
| AF218251.1 | - | 12 - 13 | 1470 | 107 | ||
| AK029316.1 | - | 12 - 13 | 3401 | 107 | ||
| AK158786.1 | - | 12 - 13 | 1437 | 107 | ||
| AK159114.1 | - | 13 - 14 | 1583 | 107 | ||
| AK172078.1 | - | 13 - 14 | 1577 | 107 | ||
| BC066839.1 | - | 13 - 14 | 1612 | 30101340 | 107 | |
| BC071182.1 | - | 13 - 14 | 1612 | 30101184 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0a1
Genomic Map