Assay Details
| Gene Symbol: Atp6v0a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243049.1 | NP_001229978.1 | 13 - 14 | 1563 | 59 | |
| NM_001243050.1 | NP_001229979.1 | 14 - 15 | 1741 | 59 | ||
| NM_001243051.1 | NP_001229980.1 | 14 - 15 | 1762 | 59 | ||
| NM_016920.3 | NP_058616.1 | 14 - 15 | 1741 | 59 | ||
| GenBank mRNA | AB022321.1 | - | 13 - 14 | 1570 | 59 | |
| AF218249.1 | - | 14 - 15 | 1711 | 59 | ||
| AF218250.1 | - | 13 - 14 | 1585 | 59 | ||
| AF218251.1 | - | 13 - 14 | 1564 | 59 | ||
| AK029316.1 | - | 13 - 14 | 3495 | 59 | ||
| AK158786.1 | - | 13 - 14 | 1531 | 59 | ||
| AK159114.1 | - | 14 - 15 | 1677 | 59 | ||
| AK166356.1 | - | 1 - 2 | 67 | 59 | ||
| AK172078.1 | - | 14 - 15 | 1671 | 59 | ||
| BC066839.1 | - | 14 - 15 | 1706 | 30101340 | 59 | |
| BC071182.1 | - | 14 - 15 | 1706 | 30101184 | 59 | |
| U13836.1 | - | 13 - 14 | 1603 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0a1
Genomic Map