Assay Details
| Gene Symbol: Il22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016971.2 | NP_058667.1 | 1 - 2 | 215 | 86 | |
| GenBank mRNA | AJ249491.1 | - | 1 - 2 | 241 | 86 | |
| BC104348.1 | - | 1 - 2 | 215 | 40049545 | 86 | |
| BC104349.1 | - | 1 - 2 | 215 | 40049546 | 86 | |
| BC116235.2 | - | 1 - 2 | 211 | 40091712 | 86 | |
| Gene Symbol: Iltifb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ249492.1 | - | 1 - 2 | 237 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Il22
Genomic Map