Assay Details
| Gene Symbol: Slc9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016981.2 | NP_058677.1 | 3 - 4 | 1854 | 103 | |
| GenBank mRNA | AK150135.1 | - | 3 - 4 | 1857 | 103 | |
| AK154857.1 | - | 3 - 4 | 1891 | 103 | ||
| AK165501.1 | - | 3 - 4 | 1854 | 103 | ||
| BC051431.1 | - | 3 - 4 | 1201 | 6529306 | 103 | |
| BC052708.1 | - | 3 - 4 | 1736 | 5702902 | 103 | |
| U51112.1 | - | 3 - 4 | 1397 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a1
Genomic Map