Assay Details
| Gene Symbol: Slc9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016981.2 | NP_058677.1 | 10 - 11 | 2815 | 83 | |
| GenBank mRNA | AK150135.1 | - | 10 - 11 | 2818 | 83 | |
| AK154857.1 | - | 10 - 11 | 2852 | 83 | ||
| AK162209.1 | - | 2 - 3 | 146 | 83 | ||
| BC051431.1 | - | 10 - 11 | 2162 | 6529306 | 83 | |
| BC052708.1 | - | 10 - 11 | 2697 | 5702902 | 83 | |
| U51112.1 | - | 10 - 11 | 2358 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a1
Genomic Map