Assay Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017366.3 | NP_059062.1 | 2 - 3 | 247 | 65 | |
| GenBank mRNA | AK076037.1 | - | 2 - 3 | 208 | 65 | |
| AU067294.1 | - | 2 - 3 | 247 | 65 | ||
| BC026559.1 | - | 2 - 3 | 226 | 5037878 | 65 | |
| Z71189.1 | - | 2 - 3 | 168 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acadvl
Genomic Map