Assay Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017366.3 | NP_059062.1 | 6 - 7 | 588 | 62 | |
| GenBank mRNA | AF017176.1 | - | 3 - 4 | 236 | 62 | |
| AK076037.1 | - | 6 - 7 | 549 | 62 | ||
| AK146334.1 | - | 1 - 2 | 61 | 62 | ||
| AK196376.1 | - | 1 - 2 | 151 | 62 | ||
| AK215457.1 | - | 1 - 2 | 151 | 62 | ||
| BC026559.1 | - | 6 - 7 | 567 | 5037878 | 62 | |
| Z71189.1 | - | 6 - 7 | 509 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acadvl
Genomic Map