Assay Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017366.3 | NP_059062.1 | 7 - 8 | 732 | 87 | |
| GenBank mRNA | AF017176.1 | - | 4 - 5 | 380 | 87 | |
| AK076037.1 | - | 7 - 8 | 693 | 87 | ||
| AK146334.1 | - | 2 - 3 | 205 | 87 | ||
| BC026559.1 | - | 7 - 8 | 711 | 5037878 | 87 | |
| Z71189.1 | - | 7 - 8 | 653 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acadvl
Genomic Map