Assay Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017366.3 | NP_059062.1 | 8 - 9 | 866 | 85 | |
| GenBank mRNA | AF017176.1 | - | 5 - 6 | 514 | 85 | |
| AK076037.1 | - | 8 - 9 | 827 | 85 | ||
| AK146334.1 | - | 3 - 4 | 339 | 85 | ||
| AK211026.1 | - | 1 - 2 | 85 | |||
| BC026559.1 | - | 8 - 9 | 5037878 | 85 | ||
| Z71189.1 | - | 8 - 9 | 787 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acadvl
Genomic Map