Assay Details
| Gene Symbol: Arhgef7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113517.1 | NP_001106989.1 | 9 - 10 | 1223 | 75 | |
| NM_001113518.1 | NP_001106990.1 | 8 - 9 | 1012 | 75 | ||
| NM_017402.4 | NP_059098.2 | 8 - 9 | 1012 | 75 | ||
| GenBank mRNA | AF247654.1 | - | 8 - 9 | 908 | 75 | |
| AF247655.1 | - | 6 - 7 | 629 | 75 | ||
| AF343877.1 | - | 8 - 9 | 892 | 75 | ||
| AK052545.1 | - | 8 - 9 | 983 | 75 | ||
| AK129064.1 | - | 9 - 10 | 1159 | 75 | ||
| AK139156.1 | - | 9 - 10 | 1152 | 75 | ||
| AY220301.1 | - | 8 - 9 | 977 | 75 | ||
| BC044838.1 | - | 8 - 9 | 997 | 3499604 | 75 | |
| U96634.1 | - | 6 - 7 | 702 | 75 | ||
More Information
Set Membership: |
|

Arhgef7
Genomic Map