Assay Details
| Gene Symbol: Atf6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017406.4 | NP_059102.2 | 14 - 15 | 1690 | 67 | |
| GenBank mRNA | AK090378.1 | - | 14 - 15 | 1640 | 67 | |
| AK143765.1 | - | 14 - 15 | 1623 | 67 | ||
| AK146671.1 | - | 14 - 15 | 1657 | 67 | ||
| BC013534.1 | - | 14 - 15 | 1690 | 4239106 | 67 | |
| BC052635.1 | - | 14 - 15 | 1625 | 6395626 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atf6b
Genomic Map