Assay Details
| Gene Symbol: Atf6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017406.4 | NP_059102.2 | 15 - 16 | 1764 | 130 | |
| GenBank mRNA | AK090378.1 | - | 15 - 16 | 1714 | 130 | |
| AK143765.1 | - | 15 - 16 | 1697 | 130 | ||
| AK146671.1 | - | 15 - 16 | 1731 | 130 | ||
| AK159169.1 | - | 15 - 16 | 1688 | 130 | ||
| BC013534.1 | - | 15 - 16 | 1764 | 4239106 | 130 | |
| BC052635.1 | - | 15 - 16 | 1699 | 6395626 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atf6b
Genomic Map