Assay Details
| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017478.3 | NP_059506.1 | 425 | 83 | ||
| GenBank mRNA | AF205065.1 | - | 328 | 83 | ||
| AJ251067.1 | - | 397 | 83 | |||
| AK079599.1 | - | 5 - 6 | 341 | 83 | ||
| AK088492.1 | - | 409 | 83 | |||
| AK147943.1 | - | 371 | 83 | |||
| AK149462.1 | - | 5 - 6 | 362 | 83 | ||
| AK163668.1 | - | 5 - 6 | 396 | 83 | ||
| BC049178.1 | - | 391 | 6309945 | 83 | ||
| BC145898.1 | - | 368 | 40131201 | 83 | ||
| BC145900.1 | - | 368 | 40131203 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Copg2
Genomic Map