Assay Details
| Gene Symbol: Tyk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205312.1 | NP_001192241.1 | 2 - 3 | 258 | 140 | |
| NM_018793.2 | NP_061263.2 | 2 - 3 | 258 | 140 | ||
| GenBank mRNA | AF173032.1 | - | 2 - 3 | 193 | 140 | |
| AK154438.1 | - | 2 - 3 | 216 | 140 | ||
| AK155923.1 | - | 2 - 3 | 171 | 140 | ||
| AK159303.1 | - | 2 - 3 | 227 | 140 | ||
| AK165483.1 | - | 2 - 3 | 227 | 140 | ||
| AK167726.1 | - | 2 - 3 | 237 | 140 | ||
| AK170107.1 | - | 2 - 3 | 197 | 140 | ||
| AK170940.1 | - | 2 - 3 | 197 | 140 | ||
| AK171609.1 | - | 2 - 3 | 171 | 140 | ||
| BC053021.1 | - | 1 - 2 | 163 | 140 | ||
| BC094240.1 | - | 1 - 2 | 154 | 30610201 | 140 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF173032.1,AK154438.1,AK159303.1,AK167726.1,AK170107.1,BC094240.1,NM_001205312.1,NM_018793.2 transcripts.
Set Membership: |
|

Tyk2
Genomic Map