Assay Details
| Gene Symbol: Tyk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205312.1 | NP_001192241.1 | 11 - 12 | 2006 | 97 | |
| NM_018793.2 | NP_061263.2 | 11 - 12 | 1937 | 97 | ||
| GenBank mRNA | AF173032.1 | - | 11 - 12 | 1872 | 97 | |
| AK154438.1 | - | 11 - 12 | 1964 | 97 | ||
| AK155923.1 | - | 11 - 12 | 1849 | 97 | ||
| AK159303.1 | - | 11 - 12 | 1906 | 97 | ||
| AK167726.1 | - | 11 - 12 | 1916 | 97 | ||
| AK170107.1 | - | 11 - 12 | 1876 | 97 | ||
| AK171609.1 | - | 11 - 12 | 1849 | 97 | ||
| BC094240.1 | - | 10 - 11 | 1833 | 30610201 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tyk2
Genomic Map