Assay Details
| Gene Symbol: Atp6ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018794.4 | NP_061264.1 | 2 - 3 | 353 | 64 | |
| GenBank mRNA | AB031290.1 | - | 2 - 3 | 271 | 64 | |
| AK093961.1 | - | 2 - 3 | 323 | 64 | ||
| AK150333.1 | - | 2 - 3 | 288 | 64 | ||
| AK158956.1 | - | 2 - 3 | 341 | 64 | ||
| AK159449.1 | - | 2 - 3 | 304 | 64 | ||
| AK159612.1 | - | 2 - 3 | 285 | 64 | ||
| AK161631.1 | - | 2 - 3 | 292 | 64 | ||
| AK166793.1 | - | 2 - 3 | 287 | 64 | ||
| AK181118.1 | - | 1 - 2 | 130 | 64 | ||
| AK216062.1 | - | 1 - 2 | 130 | 64 | ||
| AK220247.1 | - | 2 - 3 | 344 | 64 | ||
| BC048241.1 | - | 2 - 3 | 305 | 6493219 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6ap1
Genomic Map