Assay Details
| Gene Symbol: Slc1a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018861.3 | NP_061349.3 | 4 - 5 | 1037 | 114 | |
| GenBank mRNA | AF246129.1 | - | 4 - 5 | 796 | 114 | |
| AK044280.1 | - | 4 - 5 | 1008 | 114 | ||
| AK136900.1 | - | 4 - 5 | 1037 | 114 | ||
| AK139248.1 | - | 4 - 5 | 1036 | 114 | ||
| AK140866.1 | - | 4 - 5 | 1038 | 114 | ||
| AK148075.1 | - | 4 - 5 | 1038 | 114 | ||
| AK202085.1 | - | 2 - 3 | 179 | 114 | ||
| AK213050.1 | - | 2 - 3 | 179 | 114 | ||
| BC043483.1 | - | 4 - 5 | 995 | 5003744 | 114 | |
| BC052733.1 | - | 4 - 5 | 911 | 6816689 | 114 | |
| U75215.1 | - | 4 - 5 | 907 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a4
Genomic Map