Assay Details
| Gene Symbol: Cxcl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018866.2 | NP_061354.1 | 2 - 3 | 227 | 73 | |
| GenBank mRNA | AF030636.1 | - | 2 - 3 | 223 | 73 | |
| AF044196.1 | - | 2 - 3 | 227 | 73 | ||
| AK156025.1 | - | 2 - 3 | 241 | 73 | ||
| AK160241.1 | - | 2 - 3 | 243 | 73 | ||
| BC012965.1 | - | 2 - 3 | 226 | 4217539 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cxcl13
Genomic Map