Assay Details
| Gene Symbol: Tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290800.1 | NP_001277729.1 | 10 - 11 | 2090 | 61 | |
| NM_019392.2 | NP_062265.2 | 10 - 11 | 1626 | 61 | ||
| GenBank mRNA | AB000828.1 | - | 10 - 11 | 1363 | 61 | |
| AK134607.1 | - | 2 - 3 | 463 | 61 | ||
| AK141198.1 | - | 10 - 11 | 1397 | 61 | ||
| BC066058.1 | - | 10 - 11 | 1626 | 6834015 | 61 | |
| BC082325.1 | - | 10 - 11 | 1574 | 30356080 | 61 | |
| D17393.1 | - | 10 - 11 | 2068 | 61 | ||
| U05683.1 | - | 10 - 11 | 1411 | 61 | ||
| U18342.1 | - | 10 - 11 | 1543 | 61 | ||
| U18343.1 | - | 10 - 11 | 2086 | 61 | ||
| U18933.1 | - | 10 - 11 | 1562 | 61 | ||
| X78103.1 | - | 10 - 11 | 1570 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tyro3
Genomic Map