Assay Details
| Gene Symbol: Tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290800.1 | NP_001277729.1 | 16 - 17 | 2700 | 65 | |
| NM_019392.2 | NP_062265.2 | 16 - 17 | 2236 | 65 | ||
| GenBank mRNA | AB000828.1 | - | 16 - 17 | 1973 | 65 | |
| AK141198.1 | - | 16 - 17 | 2007 | 65 | ||
| BC066058.1 | - | 16 - 17 | 2236 | 6834015 | 65 | |
| BC082325.1 | - | 16 - 17 | 2184 | 30356080 | 65 | |
| D17393.1 | - | 16 - 17 | 2678 | 65 | ||
| U05683.1 | - | 16 - 17 | 2021 | 65 | ||
| U18342.1 | - | 16 - 17 | 2153 | 65 | ||
| U18343.1 | - | 16 - 17 | 2696 | 65 | ||
| U18933.1 | - | 16 - 17 | 2172 | 65 | ||
| X78103.1 | - | 16 - 17 | 2180 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tyro3
Genomic Map