Assay Details
| Gene Symbol: Tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290800.1 | NP_001277729.1 | 17 - 18 | 2859 | 78 | |
| NM_019392.2 | NP_062265.2 | 17 - 18 | 2395 | 78 | ||
| GenBank mRNA | AB000828.1 | - | 17 - 18 | 2132 | 78 | |
| AK141198.1 | - | 17 - 18 | 2166 | 78 | ||
| AK213086.1 | - | 1 - 2 | 104 | 78 | ||
| BC066058.1 | - | 17 - 18 | 2395 | 6834015 | 78 | |
| BC082325.1 | - | 17 - 18 | 2343 | 30356080 | 78 | |
| D17393.1 | - | 17 - 18 | 2837 | 78 | ||
| U05683.1 | - | 17 - 18 | 2180 | 78 | ||
| U18342.1 | - | 17 - 18 | 2312 | 78 | ||
| U18343.1 | - | 17 - 18 | 2855 | 78 | ||
| U18933.1 | - | 17 - 18 | 2331 | 78 | ||
| X78103.1 | - | 17 - 18 | 2339 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tyro3
Genomic Map