Assay Details
| Gene Symbol: Tnfsf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019418.3 | NP_062291.1 | 3 - 4 | 409 | 68 | |
| GenBank mRNA | AB029155.1 | - | 3 - 4 | 420 | 68 | |
| AF123385.1 | - | 3 - 4 | 297 | 68 | ||
| AF227533.1 | - | 3 - 4 | 307 | 68 | ||
| BC125477.1 | - | 3 - 4 | 386 | 40129992 | 68 | |
| BC125479.1 | - | 3 - 4 | 386 | 40129994 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
immune response
release of cytoplasmic sequestered NF-kappaB
positive regulation of T cell chemotaxis
T cell costimulation
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of myoblast differentiation
positive regulation of myoblast fusion
receptor binding
cytokine activity
tumor necrosis factor receptor binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
immune response
release of cytoplasmic sequestered NF-kappaB
positive regulation of T cell chemotaxis
T cell costimulation
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of myoblast differentiation
positive regulation of myoblast fusion
receptor binding
cytokine activity
tumor necrosis factor receptor binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208240 | BC125477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208240 | BC125479.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208240 | NM_019418.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208240 | AB029155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208240 | BC125477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208240 | BC125479.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208240 | NM_019418.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208240 | AB029155.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115525 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 553336 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138409 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208240 | BC125477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208240 | BC125479.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208240 | NM_019418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208240 | AB029155.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208240 | BC125477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208240 | BC125479.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208240 | NM_019418.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208240 | AB029155.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P208240 | BC125477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P208240 | BC125479.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P208240 | NM_019418.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P208240 | AB029155.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P208240 | BC125477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P208240 | BC125479.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P208240 | NM_019418.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P208240 | AB029155.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260407 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138409 | - | - |
| Whole Mouse Genome G4122A | A_51_P208240 | BC125477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208240 | BC125479.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208240 | NM_019418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208240 | AB029155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208240 | BC125477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208240 | BC125479.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208240 | NM_019418.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208240 | AB029155.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100213 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138409 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13159873 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13159874 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13070811 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13070807 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13070813 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13159875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13072749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13070812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13301559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13339331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13245753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13569602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13226781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13569604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13070811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13530655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260383 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10452313 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452308 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452310 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452312 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452311 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452315 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452314 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10452309 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450298_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421588_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10452307 | - | - |
| Mouse Genome 430 2.0 Array | 1421588_at | - | - |
| Mouse Genome 430 2.0 Array | 1450298_at | - | - |
| Mouse Genome 430 2.0 Array | 1450298_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421588_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50930_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50930_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700022064.mm.1 | NM_019418.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700022064.mm.1 | TC1700002351.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700022063.mm.1 | NM_019418.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700022063.mm.1 | TC1700002351.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421588_PM_at | - | - |
| Mouse Expression Set 430 | 1450298_PM_at | - | - |
| Mouse Expression Set 430 | 1421588_at | - | - |
| Mouse Expression Set 430 | 1450298_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450298_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421588_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421588_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450298_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450298_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450298_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421588_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421588_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 498462 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5550594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5550594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1450369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550594 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450369 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223954 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2510048 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223954 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2510048 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001714.1_20-S | - | - |
| mouse-6 v1.1 (Illumina) | scl49742.4_443-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450369 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550594 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001714.1_20-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49742.4_443-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450369 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550594 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2851576 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223954 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2851575 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223954 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2851575 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2851576 | - | - |

Tnfsf14
Genomic Map