Assay Details
| Gene Symbol: Gabbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019439.3 | NP_062312.3 | 19 - 20 | 2635 | 64 | |
| GenBank mRNA | AF008649.1 | - | 19 - 20 | 2519 | 64 | |
| AF120255.1 | - | 14 - 15 | 1963 | 64 | ||
| AK038844.1 | - | 1675 | 64 | |||
| AK156397.1 | - | 17 - 18 | 2369 | 64 | ||
| BC054735.1 | - | 19 - 20 | 2635 | 6401243 | 64 | |
| BC056990.1 | - | 14 - 15 | 2012 | 6821442 | 64 | |
More Information
Set Membership: |
|

Gabbr1
Genomic Map