Assay Details
| Gene Symbol: Stk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019442.3 | NP_062315.1 | 2 - 3 | 263 | 101 | |
| GenBank mRNA | AF099934.1 | - | 2 - 3 | 197 | 101 | |
| AK146739.1 | - | 2 - 3 | 265 | 101 | ||
| AK168865.1 | - | 2 - 3 | 265 | 101 | ||
| AK218509.1 | - | 1 - 2 | 92 | 101 | ||
| BC022681.1 | - | 2 - 3 | 263 | 4240347 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156002 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156001 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156006 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156005 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156000 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156007 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156003 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440568 | AF099934.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440568 | AK146739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440568 | AK168865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440568 | BC022681.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440568 | NM_019442.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440568 | AF099934.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440568 | AK146739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440568 | AK168865.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440568 | BC022681.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P440568 | NM_019442.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13946 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160915 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163811 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440568 | AF099934.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440568 | AK146739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440568 | AK168865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440568 | BC022681.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440568 | NM_019442.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440568 | AF099934.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440568 | AK146739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440568 | AK168865.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440568 | BC022681.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P440568 | NM_019442.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31155993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31155987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31155977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31155997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31155989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31155999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31155991 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160915 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163811 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03310885 | - | - |
| Whole Mouse Genome G4122A | A_52_P256844 | AF099934.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P256844 | AK146739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P256844 | AK168865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P256844 | BC022681.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P256844 | NM_019442.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440568 | AF099934.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440568 | AK146739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440568 | AK168865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440568 | BC022681.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440568 | NM_019442.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440568 | AF099934.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440568 | AK146739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440568 | AK168865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440568 | BC022681.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P440568 | NM_019442.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P254607 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163811 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160915 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13174870 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13537998 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156000 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156002 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156007 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156005 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156006 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156001 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31155934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31155999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31155997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31155988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13525714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31155968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31155997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31155992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31155988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13537998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31155991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31155994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31155985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13559943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31155993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31155987 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450270 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450267 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450271 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450268 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450269 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417334_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450265 | - | - |
| Mouse Genome 430 2.0 Array | 1417334_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417334_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700018143.mm.1 | NM_019442.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018143.mm.1 | NM_019442.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018143.mm.1 | TC1700001928.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018145.mm.1 | NM_019442.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018145.mm.1 | NM_019442.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018145.mm.1 | TC1700001928.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018144.mm.1 | NM_019442.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018144.mm.1 | NM_019442.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018144.mm.1 | TC1700001928.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417334_PM_at | - | - |
| Mouse Expression Set 430 | 1417334_at | - | - |
| Murine Genome U74v2 Set | 98456_at | - | - |
| Murine Genome U74v2 Set | 98456_PM_at | - | - |
| Murine Genome U74v2 Set | 170329_PM_at | - | - |
| Murine Genome U74v2 Set | 170329_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417334_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417334_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417334_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417334_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 803714 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6660136 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6660136 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6620300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3120070 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003120070 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006620300 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660136 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2649355 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232972 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2751865 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2751865 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2649355 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232972 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001620.1_1-S | - | - |
| mouse-6 v1.1 (Illumina) | scl50012.2.189_28-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001697.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3120070 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6620300 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660136 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001620.1_1-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001697.1_9-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50012.2.189_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3120070 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660136 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620300 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2751865 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2649355 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1232972 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2751865 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2649355 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232972 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50012.2.189_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001620.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001697.1_9-S | - | - |

Stk19
Genomic Map