Assay Details
| Gene Symbol: Stk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019442.3 | NP_062315.1 | 5 - 6 | 602 | 60 | |
| GenBank mRNA | AF099934.1 | - | 5 - 6 | 536 | 60 | |
| AK146739.1 | - | 5 - 6 | 604 | 60 | ||
| AK168865.1 | - | 5 - 6 | 604 | 60 | ||
| BC022681.1 | - | 5 - 6 | 602 | 4240347 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stk19
Genomic Map