Assay Details
| Gene Symbol: Uso1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019490.1 | NP_062363.1 | 7 - 8 | 718 | 97 | |
| GenBank mRNA | AF096868.1 | - | 6 - 7 | 506 | 97 | |
| AK144638.1 | - | 7 - 8 | 709 | 97 | ||
| AK156070.1 | - | 7 - 8 | 711 | 97 | ||
| AK166587.1 | - | 8 - 9 | 678 | 97 | ||
| AK172425.1 | - | 7 - 8 | 709 | 97 | ||
| BC016069.1 | - | 7 - 8 | 718 | 4480992 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uso1
Genomic Map