Assay Details
| Gene Symbol: Ccl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019577.4 | NP_062523.1 | 1 - 2 | 142 | 63 | |
| GenBank mRNA | AF244367.1 | - | 1 - 2 | 71 | 63 | |
| AF281075.1 | - | 1 - 2 | 124 | 63 | ||
| AK009718.1 | - | 1 - 2 | 114 | 63 | ||
| AK157068.1 | - | 1 - 2 | 143 | 63 | ||
| BC065389.1 | - | 1 - 2 | 148 | 3472002 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccl24
Genomic Map